Diisobutylaluminium hydride 20% (w/w) 1.2 M in toluene, AcroSeal®
Supplier: Thermo Scientific
Danger
Synonyms: Di-i-butylaluminum hydride, DIBAL-H
201081000 201084000 201088000
201081000.EA 59.4 EUR
201081000. 201084000. 201088000.
Diisobutylaluminium hydride 20% (w/w) 1.2 M in toluene, AcroSeal®
Diisobutylaluminium hydride
Formula: C₈H₁₉Al
MW: 142.22 g/mol
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00008928
CAS Number: 1191-15-7
EINECS: 214-729-9
UN: 3399
ADR: 4.3,I
Order Now
Specification Test Results
Appearance Clear solution
Strength 19.5 to 20.5 % (DIBAL-H, on Al basis)