Trichloroacetic acid (TCA) 24% in aqueous solution
Supplier: PROLABO CMP
PRLS88986.360EA 0 EUR
PRLS88986.360
Trichloroacetic acid (TCA) 24% in aqueous solution
Trichloroacetic acid (TCA)
Formula: Cl₃CCOOH
MW: 163,39 g/mol
Boiling Pt: 196 °C (1013 hPa)
Melting Pt: 54…56 °C
Density: ≤1,07 g/cm³ (20 °C)
MDL Number: MFCD00004177
CAS Number: 76-03-9
UN: 2564
ADR: 8,II
https://si.vwr.com/stibo/web/std.lang.all/74/52/727452.jpg