Sulphuric acid 0.015 mol/l (0.03 N) in aqueous solution
Supplier: Hach
2620532 2620553
HACH2620532EA 23.6 EUR
HACH2620532 HACH2620553
Sulphuric acid 0.015 mol/l (0.03 N) in aqueous solution
Sulphuric acid
Formula: H₂SO₄
Density: 1.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8,II