Sulphuric acid 2.625 mol/l (5.25 N) in aqueous solution
Supplier: Hach
Danger
244953 244903 244932
HACH244953EA 38.1 EUR
HACH244953 HACH244903 HACH244932
Sulphuric acid 2.625 mol/l (5.25 N) in aqueous solution
Sulphuric acid
Formula: H₂SO₄
Density: 1.146 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8,II