Single channel pipettes, mechanical, fixed volume, Pipet-Lite™
Supplier: RAININ
FSL-25 FSL-20
RAINFSL-25EA 0 EUR
RAINFSL-25 RAINFSL-20
Single channel pipettes, mechanical, fixed volume, Pipet-Lite™
Pipettes Micropipettes
This light-weight, ergonomically shaped pipette with fingerhook is ideal for dispensing large number of samples or reagents with dedicated volumes.
  • Shaft and tip-ejector autoclavable
  • Low force plunging
  • Silicone shock absorber: Extra thumb protection
  • Easy volume adjustment including volume lock
  • Quick-release tip ejector