Tetraethylammonium toluene-4-sulphonate, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Tetraethylammonium p-toluenesulphonate, Tetraethylammonium p-Toluenesulphonate
M-Clarity™ quality level = MQ100
134473-100G
134473-500G
SIAL134473-100GEA
148
EUR
SIAL134473-100G
SIAL134473-500G
Tetraethylammonium toluene-4-sulphonate, Sigma-Aldrich®
Tetraethylammonium toluene-4-sulphonate
Formula:
C₁₅H₂₇NO₃S MW: 301.4500 g/mol |
MDL Number:
MFCD00011831 CAS Number: 733-44-8 EINECS: 211-992-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...