Bis(benzonitrile)palladium(II) dichloride, Sigma-Aldrich®

Supplier: Merck

Synonyms: Bis(benzonitrile)palladium chloride, Dichlorobis(benzonitrile)palladium(II), Bis(benzonitrile)palladium(II) Dichloride

M-Clarity™ quality level = MQ100

223689-10G 223689-1G
SIAL223689-10GEA 700 EUR
SIAL223689-10G SIAL223689-1G
Bis(benzonitrile)palladium(II) dichloride, Sigma-Aldrich®
Bis(benzonitrile)dichloropalladium(II)
Formula: C₁₄H₁₀Cl₂N₂Pd
MDL Number: MFCD00013123
CAS Number: 14220-64-5
EINECS: 238-085-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR