2-Chloro-5-(tributylstannyl)pyrimidine, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ100

706868-1G 706868-500MG
SIAL706868-1GEA 266 EUR
SIAL706868-1G SIAL706868-500MG
2-Chloro-5-(tributylstannyl)pyrimidine, Sigma-Aldrich®
2-Chloro-5-(tributylstannyl)pyrimidine
Formula: C₁₆H₂₉ClN₂Sn
MDL Number: MFCD07787378
CAS Number: 155191-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR