1-(3-Chloro-5-(trifluoromethyl)-2-pyridinyl)-1,4-diazepane ≥98%
Supplier: Thermo Scientific
Danger
Synonyms: 1-(3-Chloro-5-trifluoromethyl-2-pyridyl)homopiperazine
H50263.03 H50263.06
H50263.03EA 112 EUR
H50263.03 H50263.06
1-(3-Chloro-5-(trifluoromethyl)-2-pyridinyl)-1,4-diazepane ≥98%
1-(3-Chloro-5-(trifluoromethyl)-2-pyridinyl)-1,4-diazepane
Purity: 98% CAS No.: 231953-40-5 Molecular Formula: C11H13ClF3N3 Formula Weight: 279.69 Boiling Point: 128°/1mm UN No.: 2735 MDL No.: MFCD00238856
Formula: C₁₁H₁₃ClF₃N₃
MW: 279.69 g/mol
Boiling Pt: 128 °C (1 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00238856
CAS Number: 231953-40-5
ADR: 8,III