Choline hydroxide 46% (w/w) in aqueous solution
Supplier: Thermo Scientific
Danger
Synonyms: 2-Hydroxyethyltrimethylammonium Hydroxide, Choline
B24443.AC B24443.AE
B24443.ACEA 35.3 EUR
B24443.AC B24443.AE
Choline hydroxide 46% (w/w) in aqueous solution
Choline hydroxide
CAS No.: 123-41-1 Molecular Formula: C5H15NO2 Formula Weight: 121.18 Refractive Index: 1.4270 UN No.: 3267 MDL No.: MFCD00002831
Formula: C₅H₁₅NO₂
MW: 121.18 g/mol
Density: 1.073
Storage Temperature: Ambient
MDL Number: MFCD00002831
CAS Number: 123-41-1
EINECS: 204-625-1
UN: 3267
ADR: 8,II