1-(4-Hydroxyphenyl)-5-mercapto-1H-tetrazole 96%
Supplier: Thermo Scientific
Warning
Synonyms: 1-(4-Hydroxyphenyl)-5-mercaptotetrazole
B22003.06 B22003.14 B22003.22
B22003.06EA 27.3 EUR
B22003.06 B22003.14 B22003.22
1-(4-Hydroxyphenyl)-5-mercapto-1H-tetrazole 96%
1-(4-Hydroxyphenyl)-5-mercapto-1H-tetrazole
Purity: 96% CAS No.: 52431-78-4 Molecular Formula: C7H6N4OS Formula Weight: 194.21 Melting Point: 169-171° MDL No.: MFCD00132898
Formula: C₇H₆N₄OS
MW: 194.21 g/mol
Melting Pt: 169…171 °C
Storage Temperature: Ambient
MDL Number: MFCD00132898
CAS Number: 52431-78-4
EINECS: 257-909-2