Ethyl 4-hydroxy-6-(trifluoromethoxy)quinoline-3-carboxylate

Supplier: Thermo Scientific

Synonyms: Ethyl 4-hydroxy-6-(trifluoromethoxy)quinoline-3-carboxylate

KM09296.1 KM09296.10
MAYBKM09296.1EA 0 EUR
MAYBKM09296.1 MAYBKM09296.10
Ethyl 4-hydroxy-6-(trifluoromethoxy)quinoline-3-carboxylate
Ethyl 4-hydroxy-6-(trifluoromethoxy)quinoline-3-carboxylate
Formula: C₁₃H₁₀F₃NO₄
MW: 301.2200 g/mol
MDL Number: MFCD00219847
CAS Number: 175203-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR