4-Cyano-4'-pentylbiphenyl 99
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 4'-Pentyl-4-biphenylcarbonitrile, 4'-Pentyl-[1,1'-biphenyl]-4-carbonitrile
B21856.03 B21856.06
B21856.03EA 66.6 EUR
B21856.03 B21856.06
4-Cyano-4'-pentylbiphenyl 99
4-Cyano-4'-pentylbiphenyl
Purity: 99% CAS No.: 40817-08-1 Molecular Formula: C18H19N Formula Weight: 249.36 Flash Point: 110°(230°F) Refractive Index: 1.5330 Melting Point: 22-23° Boiling Point: 212-215°/3mm UN No.: 3276 MDL No.: MFCD00036350
Formula: C₁₈H₁₉N
MW: 249.36 g/mol
Boiling Pt: 212…215 °C (3 mmHg)
Melting Pt: 22…23 °C
Density: 1.008
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00036350
CAS Number: 40817-08-1
EINECS: 255-093-2
ADR: 6.1 III