3-Amino-6-chloro-4-methylpyridine 98%
Supplier: Thermo Scientific
Warning
Synonyms: 3-Amino-6-chloro-4-picoline, 5-Amino-2-chloro-4-methylpyridine, 6-Chloro-4-methylpyridin-3-amine
360990010 360990050
ACRO360990010EA 79.1 EUR
ACRO360990010 ACRO360990050
3-Amino-6-chloro-4-methylpyridine 98%
3-Amino-6-chloro-4-methylpyridine
Formula: C₆H₇ClN₂
MW: 142.59 g/mol
Melting Pt: 69…73 °C
Storage Temperature: Ambient
MDL Number: MFCD03427661
CAS Number: 66909-38-4
UN: 2811
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Bordeaux to brown Powder
Infrared spectrum Conforms
Melting point 69 to 73 °C
GC ≥97.5 %