3,4-Dimethyl-2-hexanol erythro + threo ≥98%
Supplier: Thermo Scientific
Warning
B20243.03 B20243.14
B20243.03EA 36.9 EUR
B20243.03 B20243.14
3,4-Dimethyl-2-hexanol erythro + threo ≥98%
3,4-Dimethyl-2-hexanol
Purity: 98% CAS No.: 31350-88-6 Molecular Formula: C8H18O Formula Weight: 130.23 MDL No.: MFCD00021887
Formula: C₈H₁₈O
MW: 130.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00021887
CAS Number: 31350-88-6