Potassium dichromate 1/6 mol/l (1 N) in aqueous solution

Supplier: Hach

HACH2197153EA 65.6 EUR
HACH2197153
Potassium dichromate 1/6 mol/l (1 N) in aqueous solution
Potassium dichromate
Formula: K₂Cr₂O₇
MW: 294,18 g/mol
MDL Number: MFCD00011367
CAS Number: 7778-50-9
UN: 3287
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR