(Phenylthio)acetonitrile

Supplier: Thermo Scientific

Synonyms: 2-(Phenylthio)acetonitrile

TL00082.1 TL00082.10 TL00082.50
MAYBTL00082.1EA 0 EUR
MAYBTL00082.1 MAYBTL00082.10 MAYBTL00082.50
(Phenylthio)acetonitrile
(Phenylthio)acetonitrile
Formula: C₈H₇NS
MW: 149.22 g/mol
MDL Number: MFCD00019831
CAS Number: 5219-61-4
EINECS: 226-013-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR