VWR®, Polynuclear Aromatic Hydrocarbons (PAH) Mix 3
Supplier: Avantor
VWR®
124832SEA 129 EUR
124832S
VWR®, Polynuclear Aromatic Hydrocarbons (PAH) Mix 3
Standards Environmental Analysis Standards
16 components
Five μg/ml : Benzo(k)fluoranthene
10 μg/ml : Benzo(a)pyrene; Dibenz(a,h)anthracene; Fluoranthene; Benz(a)anthracene; Benzo(b)fluoranthene; Benzo(g,h,i)perylene; Indeno(1,2,3-cd)pyrene; Chrysene; Pyrene
100 μg/ml : Fluorene; Naphthalene; Acenaphthylene; Anthracene; Phenanthrene; Acenaphthene