Storage cabinets for gas cylinders, GFL-F90
Supplier: LACONT UMWELTTECHNIK
C66-2012-A C66-2014-A C66-2015-A C66-2001-A C66-2002-A C66-2004-A C66-2000-A C66-2010-A
LACOC66-2012-AEA 0 EUR
LACOC66-2012-A LACOC66-2014-A LACOC66-2015-A LACOC66-2001-A LACOC66-2002-A LACOC66-2004-A LACOC66-2000-A LACOC66-2010-A
Storage cabinets for gas cylinders, GFL-F90
Furniture Storage Cabinets
Gas cylinder storage system for regular storage outdoors of pressurised gas cylinders.
  • Roof and walls in fire-resistant version
  • Cantilever arm compartment divider with safety chains (fold out)
  • Lockable grilled door
  • Mounted delivery
  • With hinged or double-hinged door depending on version
Certifikace: Conforms with TRGS 510, DIN 4102.