Shelves for drums with sump pallets, FR
Supplier: LACONT UMWELTTECHNIK
R74-1023-A R74-1022-A R74-1024-A R74-1025-A R74-1006-A R74-1007-A R74-1008-A R74-1009-A R74-1010-A R74-1011-A R74-1012-A R74-1013-A
LACOR74-1023-AEA 0 EUR
LACOR74-1023-A LACOR74-1022-A LACOR74-1024-A LACOR74-1025-A LACOR74-1006-A LACOR74-1007-A LACOR74-1008-A LACOR74-1009-A LACOR74-1010-A LACOR74-1011-A LACOR74-1012-A LACOR74-1013-A
Shelves for drums with sump pallets, FR
Furniture Shelves
Shelves are used for horizontal storage and filling of various container sizes.
  • Collecting trays made of 3 mm sheet steel
  • Longitudinal bars height adjustable
  • High stability through plug and screw connections
  • Basic and cultivation fields can be combined with each other
  • Frame parts galvanised - longitudinal beams powder-coated
Capacity per truss pair: 600 kg.
Drum racks with sump capacity 210 L and sump dimension W×D×H: 1325×1320×250 mm.