Trousers for men, C.I., Design C, royal blue
Supplier: Fristads Kansas Group
KWIN843530539C58EA 0 EUR
KWIN843530539C58 KWIN843530210C152 KWIN843530210C154 KWIN843530539C56 KWIN843530539C54 KWIN843530210C156 KWIN843530539C52 KWIN843530520C150 KWIN843530640C54 KWIN843530210C56 KWIN843530941C46 KWIN843530520C152 KWIN843530640C56 KWIN843530941C44 KWIN843530210C54 KWIN843530640C50 KWIN843530520C154 KWIN843530210C52 KWIN843530520C156 KWIN843530210C50 KWIN843530941C48 KWIN843530640C52 KWIN843530640C148 KWIN843530440C44 KWIN843530640C48 KWIN843530210C48 KWIN843530331C44 KWIN843530210C46 KWIN843530539C50 KWIN843530520C46 KWIN843530640C146 KWIN843530210C150 KWIN843530520C44 KWIN843530940C148 KWIN843530539C46 KWIN843530539C44 KWIN843530940C146 KWIN843530210C146 KWIN843530941C52 KWIN843530210C148 KWIN843530941C50 KWIN843530331C46 KWIN843530440C48 KWIN843530210C44 KWIN843530941C56 KWIN843530640C44 KWIN843530440C46 KWIN843530331C48 KWIN843530640C46 KWIN843530941C54 KWIN843530231C44 KWIN843530539C48 KWIN843530941C58 KWIN843530331C50 KWIN843530440C50 KWIN843530331C52 KWIN843530331C54 KWIN843530440C54 KWIN843530331C56 KWIN843530440C52 KWIN843530940C156 KWIN843530231C150 KWIN843530940C52 KWIN843530940C50 KWIN843530231C152 KWIN843530530C60 KWIN843530940C56 KWIN843530530C62 KWIN843530331C58 KWIN843530940C150 KWIN843530940C54 KWIN843530940C154 KWIN843530940C58 KWIN843530940C152 KWIN843530331C60 KWIN843530131C146 KWIN843530530C58 KWIN843530331C62 KWIN843530539C146 KWIN843530530C148 KWIN843530131C148 KWIN843530231C154 KWIN843530231C156 KWIN843530530C146 KWIN843530131C54 KWIN843530530C150 KWIN843530131C52 KWIN843530640C60 KWIN843530131C50 KWIN843530940C62 KWIN843530940C60 KWIN843530440C146 KWIN843530530C50 KWIN843530440C148 KWIN843530530C52 KWIN843530131C152 KWIN843530640C62 KWIN843530530C54 KWIN843530539C148 KWIN843530210C62 KWIN843530131C150 KWIN843530210C60 KWIN843530530C56 KWIN843530530C48 KWIN843530131C156 KWIN843530539C154 KWIN843530131C154 KWIN843530539C156 KWIN843530539C150 KWIN843530640C58 KWIN843530539C152 KWIN843530210C58 KWIN843530530C156 KWIN843530131C48 KWIN843530530C152 KWIN843530131C46 KWIN843530231C146 KWIN843530530C154 KWIN843530131C44 KWIN843530231C148 KWIN843530730C148 KWIN843530730C54 KWIN843530730C146 KWIN843530730C52 KWIN843530131C62 KWIN843530730C58 KWIN843530131C60 KWIN843530730C56 KWIN843530530C44 KWIN843530440C154 KWIN843530730C50 KWIN843530530C46 KWIN843530440C156 KWIN843530440C150 KWIN843530941C146 KWIN843530440C152 KWIN843530941C148 KWIN843530331C148 KWIN843530331C146 KWIN843530131C58 KWIN843530131C56 KWIN843530331C150 KWIN843530940C44 KWIN843530730C152 KWIN843530730C150 KWIN843530730C62 KWIN843530730C156 KWIN843530940C48 KWIN843530730C60 KWIN843530730C154 KWIN843530940C46 KWIN843530331C156 KWIN843530331C154 KWIN843530331C152 KWIN843530941C60 KWIN843530941C62 KWIN843530440C58 KWIN843530231C50 KWIN843530440C56 KWIN843530231C54 KWIN843530231C52 KWIN843530440C62 KWIN843530231C48 KWIN843530231C46 KWIN843530440C60 KWIN843530520C60 KWIN843530520C62 KWIN843530730C44 KWIN843530520C148 KWIN843530520C48 KWIN843530730C48 KWIN843530730C46 KWIN843530539C62 KWIN843530640C150 KWIN843530231C62 KWIN843530231C60 KWIN843530520C146 KWIN843530941C156 KWIN843530231C58 KWIN843530231C56 KWIN843530520C50 KWIN843530520C54 KWIN843530539C60 KWIN843530640C152 KWIN843530941C150 KWIN843530520C52 KWIN843530640C154 KWIN843530941C152 KWIN843530520C58 KWIN843530640C156 KWIN843530520C56 KWIN843530941C154
Trousers for men, C.I., Design C, royal blue
Workwear
Robust trousers made from 65% polyester and 35% cotton, with soft nap on reverse side.
  • Fly with zip
  • Two front pockets, two back pockets with flap and velcro closure
  • Leg pockets with flaps and three tool pockets, leg pocket with mobile phone pocket and two pen pockets
  • Folding-ruler pocket, D-ring, inside knee pockets
Weight: 260 g/m²
Basic colour: Royal blue
Informace pro objednávání: Trousers are available in 12 different accent colours.