Overalls with bib and braces for men, C.I., design c, true navy
Supplier: Fristads Kansas Group
KWIN870539331C152EA 0 EUR
KWIN870539331C152 KWIN870539730C156 KWIN870539331C154 KWIN870539331C156 KWIN870539730C150 KWIN870539730C154 KWIN870539730C152 KWIN870539530C54 KWIN870539941C146 KWIN870539940C48 KWIN870539530C52 KWIN870539530C58 KWIN870539940C44 KWIN870539941C148 KWIN870539530C56 KWIN870539940C46 KWIN870539530C50 KWIN870539730C148 KWIN870539730C146 KWIN870539331C146 KWIN870539331C148 KWIN870539440C156 KWIN870539440C152 KWIN870539440C154 KWIN870539530C62 KWIN870539530C60 KWIN870539331C150 KWIN870539730C50 KWIN870539730C52 KWIN870539210C146 KWIN870539440C146 KWIN870539730C58 KWIN870539210C50 KWIN870539131C150 KWIN870539440C148 KWIN870539210C148 KWIN870539210C52 KWIN870539131C152 KWIN870539730C54 KWIN870539539C148 KWIN870539210C54 KWIN870539640C48 KWIN870539730C56 KWIN870539539C146 KWIN870539210C56 KWIN870539640C46 KWIN870539131C146 KWIN870539210C58 KWIN870539640C44 KWIN870539131C148 KWIN870539440C150 KWIN870539530C148 KWIN870539940C62 KWIN870539530C146 KWIN870539231C154 KWIN870539231C152 KWIN870539210C150 KWIN870539940C60 KWIN870539231C156 KWIN870539210C154 KWIN870539730C60 KWIN870539210C152 KWIN870539730C62 KWIN870539231C150 KWIN870539940C156 KWIN870539210C156 KWIN870539940C152 KWIN870539530C48 KWIN870539940C154 KWIN870539210C44 KWIN870539210C46 KWIN870539940C150 KWIN870539210C48 KWIN870539640C58 KWIN870539539C156 KWIN870539940C58 KWIN870539539C154 KWIN870539640C56 KWIN870539940C54 KWIN870539530C46 KWIN870539640C54 KWIN870539331C60 KWIN870539131C154 KWIN870539539C152 KWIN870539530C44 KWIN870539131C156 KWIN870539640C52 KWIN870539940C56 KWIN870539539C150 KWIN870539940C50 KWIN870539640C50 KWIN870539530C156 KWIN870539530C154 KWIN870539940C52 KWIN870539331C62 KWIN870539231C148 KWIN870539530C152 KWIN870539530C150 KWIN870539231C146 KWIN870539940C148 KWIN870539520C56 KWIN870539331C58 KWIN870539520C58 KWIN870539440C58 KWIN870539520C52 KWIN870539940C146 KWIN870539520C54 KWIN870539131C44 KWIN870539520C50 KWIN870539231C46 KWIN870539231C48 KWIN870539231C44 KWIN870539640C148 KWIN870539941C56 KWIN870539640C62 KWIN870539941C58 KWIN870539941C52 KWIN870539440C54 KWIN870539640C60 KWIN870539331C52 KWIN870539131C46 KWIN870539941C54 KWIN870539440C56 KWIN870539331C50 KWIN870539131C48 KWIN870539440C50 KWIN870539331C56 KWIN870539640C146 KWIN870539941C50 KWIN870539331C54 KWIN870539440C52 KWIN870539131C50 KWIN870539331C48 KWIN870539131C52 KWIN870539440C46 KWIN870539210C60 KWIN870539131C54 KWIN870539440C48 KWIN870539520C60 KWIN870539210C62 KWIN870539730C46 KWIN870539520C152 KWIN870539520C62 KWIN870539520C150 KWIN870539730C48 KWIN870539520C156 KWIN870539520C154 KWIN870539730C44 KWIN870539941C48 KWIN870539941C44 KWIN870539941C46 KWIN870539131C56 KWIN870539131C58 KWIN870539440C44 KWIN870539331C46 KWIN870539331C44 KWIN870539520C148 KWIN870539539C50 KWIN870539131C60 KWIN870539520C146 KWIN870539131C62 KWIN870539231C60 KWIN870539231C62 KWIN870539539C48 KWIN870539539C46 KWIN870539539C44 KWIN870539520C46 KWIN870539539C62 KWIN870539520C48 KWIN870539539C60 KWIN870539520C44 KWIN870539231C58 KWIN870539231C54 KWIN870539941C156 KWIN870539231C56 KWIN870539941C154 KWIN870539231C50 KWIN870539231C52 KWIN870539640C152 KWIN870539941C62 KWIN870539539C58 KWIN870539640C150 KWIN870539539C56 KWIN870539640C156 KWIN870539941C152 KWIN870539440C60 KWIN870539539C54 KWIN870539640C154 KWIN870539941C150 KWIN870539440C62 KWIN870539941C60 KWIN870539539C52
Overalls with bib and braces for men, C.I., design c, true navy
Workwear
These overalls with bib and braces are made from 65% polyester, 35% cotton, with a soft nap on reverse side.
  • Fly with zip
  • Two chest pockets, one with flap, the other with pen pocket
  • Two front pockets, two back pockets with flap
  • Leg pocket with flap and three small pockets
  • Folding-rule pocket, leg pocket with mobile phone pocket with flap and two small pockets
  • Adjustable waist, side openings
  • Knee pockets that open from the inside and can accommodate height-adjustable kneepads
Weight: 260 g/m²
Basic colour: Navy blue
Informace pro objednávání: Available with 12 different accent colours.