Trousers for men, C.I., Design C, true navy
Supplier: Fristads Kansas Group
KWIN843539530C62EA 0 EUR
KWIN843539530C62 KWIN843539940C54 KWIN843539530C60 KWIN843539940C52 KWIN843539940C58 KWIN843539940C56 KWIN843539210C156 KWIN843539539C46 KWIN843539210C154 KWIN843539539C44 KWIN843539940C62 KWIN843539940C148 KWIN843539940C146 KWIN843539940C60 KWIN843539210C152 KWIN843539640C148 KWIN843539640C146 KWIN843539210C150 KWIN843539539C156 KWIN843539131C156 KWIN843539210C48 KWIN843539539C154 KWIN843539640C46 KWIN843539210C46 KWIN843539539C152 KWIN843539640C48 KWIN843539210C44 KWIN843539131C152 KWIN843539940C44 KWIN843539940C156 KWIN843539131C154 KWIN843539640C44 KWIN843539940C154 KWIN843539940C48 KWIN843539940C152 KWIN843539940C46 KWIN843539131C150 KWIN843539539C48 KWIN843539940C150 KWIN843539530C152 KWIN843539539C56 KWIN843539539C54 KWIN843539530C154 KWIN843539730C62 KWIN843539539C52 KWIN843539530C156 KWIN843539640C150 KWIN843539940C50 KWIN843539539C50 KWIN843539539C150 KWIN843539231C146 KWIN843539640C154 KWIN843539640C152 KWIN843539231C148 KWIN843539530C150 KWIN843539640C156 KWIN843539331C44 KWIN843539331C46 KWIN843539440C148 KWIN843539331C48 KWIN843539730C60 KWIN843539440C146 KWIN843539539C58 KWIN843539730C54 KWIN843539730C52 KWIN843539730C58 KWIN843539231C152 KWIN843539730C56 KWIN843539539C62 KWIN843539231C150 KWIN843539539C60 KWIN843539231C156 KWIN843539231C154 KWIN843539440C152 KWIN843539440C150 KWIN843539231C60 KWIN843539730C146 KWIN843539730C50 KWIN843539440C156 KWIN843539440C154 KWIN843539231C62 KWIN843539331C154 KWIN843539331C156 KWIN843539730C46 KWIN843539730C148 KWIN843539730C44 KWIN843539520C62 KWIN843539730C48 KWIN843539331C150 KWIN843539331C152 KWIN843539231C58 KWIN843539440C60 KWIN843539331C60 KWIN843539231C56 KWIN843539331C62 KWIN843539520C60 KWIN843539941C146 KWIN843539440C62 KWIN843539941C148 KWIN843539231C50 KWIN843539730C154 KWIN843539730C156 KWIN843539730C150 KWIN843539231C54 KWIN843539730C152 KWIN843539231C52 KWIN843539520C58 KWIN843539331C146 KWIN843539331C148 KWIN843539520C52 KWIN843539131C48 KWIN843539520C50 KWIN843539520C56 KWIN843539131C44 KWIN843539520C54 KWIN843539440C50 KWIN843539231C46 KWIN843539331C50 KWIN843539231C44 KWIN843539331C52 KWIN843539440C54 KWIN843539941C156 KWIN843539331C54 KWIN843539440C52 KWIN843539231C48 KWIN843539440C58 KWIN843539331C56 KWIN843539941C58 KWIN843539440C56 KWIN843539331C58 KWIN843539941C54 KWIN843539941C56 KWIN843539941C60 KWIN843539520C48 KWIN843539941C62 KWIN843539941C152 KWIN843539941C154 KWIN843539520C46 KWIN843539520C44 KWIN843539941C150 KWIN843539530C48 KWIN843539131C148 KWIN843539539C146 KWIN843539530C46 KWIN843539640C58 KWIN843539210C58 KWIN843539210C56 KWIN843539640C54 KWIN843539941C46 KWIN843539440C46 KWIN843539210C54 KWIN843539941C48 KWIN843539640C56 KWIN843539210C52 KWIN843539440C44 KWIN843539640C50 KWIN843539530C44 KWIN843539520C146 KWIN843539210C50 KWIN843539941C44 KWIN843539640C52 KWIN843539539C148 KWIN843539440C48 KWIN843539941C50 KWIN843539131C62 KWIN843539941C52 KWIN843539520C148 KWIN843539131C60 KWIN843539530C146 KWIN843539530C148 KWIN843539530C58 KWIN843539520C150 KWIN843539520C154 KWIN843539530C52 KWIN843539520C152 KWIN843539530C50 KWIN843539210C62 KWIN843539530C56 KWIN843539640C60 KWIN843539520C156 KWIN843539210C60 KWIN843539640C62 KWIN843539530C54 KWIN843539131C52 KWIN843539131C50 KWIN843539210C148 KWIN843539210C146 KWIN843539131C58 KWIN843539131C56 KWIN843539131C54
Trousers for men, C.I., Design C, true navy
Workwear
Robust trousers made from 65% polyester and 35% cotton, with soft nap on reverse side.
  • Fly with zip
  • Two front pockets, two back pockets with flap and velcro® closure
  • Leg pockets with flaps and three tool pockets, leg pocket with mobile phone pocket and two pen pockets
  • Folding-ruler pocket, D-ring, inside knee pockets
Weight: 260 g/m²
Basic colour: Navy blue
Informace pro objednávání: Trousers are available in 12 different accent colours.