Avantor® Hichrom PAH2, HPLC Columns
Supplier: VWR
Avantor® Hichrom
HI-5PAH2-10CM5EA 0 EUR
HI-5PAH2-10CM5 HI-5PAH2-250A HI-5PAH2-150AS HI-5PAH2-250AM HI5PAH210C5SK HI-5PAH2-100A HI-5PAH2-200A HI-5PAH2-250AS HI-5PAH2-10C5 HI-5PAH2-100AM HI-5PAH2-250AF
Avantor® Hichrom PAH2, HPLC Columns
Chromatography Columns
Hichrom PAH2 is based on an alkyl bonded silica material with a high carbon loading, designed specifically for the analysis of Polynuclear aromatic hydrocarbons (PAHs).
  • Optimised for EPA method 610
  • Excellent separation of 16 PAHs