VWR®, Colour standard solutions, Reag. Ph. Eur.
Supplier: VWR Chemicals
85748.180EA
119
EUR
85748.180
85749.180
85750.180
85751.180
85751.260
VWRS87169.260
VWR®, Colour standard solutions, Reag. Ph. Eur.
Standards
Colour Reference and Spectroscopy Standards
Ready to use colour solutions according to Reag. Ph. Eur.
- Ready to use solutions
- Comprehensive Certificates of Analysis for all batches
- Labelled with minimum shelf life/batch number
Informace pro objednávání: Normally manufactured to order. Please check with your local Avantor sales office.