Handheld refractometers
Supplier: Reichert
635-2117EA 1760 EUR
635-2117
Handheld refractometers
Refractometers Handheld Refractometers
High precision handheld clinical refractometer that determines urine specific gravity, protein concentration in plasma or serum and total solids in other aqueous solutions.
  • Readings with a degree of accuracy, reproducibility and speed
  • Small sample required for about 0,005 ml for accurate analysis
  • Automatic temperature compensation (ATC) using hermetically sealed, liquid-filled prism that changes index with temperature