Heat resistant gloves
Supplier: Jutec
JUTEH05LA238W2EA 181 EUR
JUTEH05LA238W2
Heat resistant gloves
Gloves
These gloves consist of a Sebatan® leather palm and an aluminium coated back. They provide protection against radiation heat up to 1000 °C and contact heat up to 100 °C.
  • Flexible, comfortable because of its soft aluminium coating
  • Abrasion resistant
Ideal for use in welding, foundries, metal industry, ship repairs, etc.
Certifikace: EN 388 (4X4X), EN 407 (413444); Cat. III