Heat resistant gloves
Supplier: Jutec
112-1963EA 133 EUR
112-1963
Heat resistant gloves
Gloves
These double insulation aramid gloves provide protection against contact heat up to 500 °C.
  • Cut resistant, abrasion resistant
  • Five-finger gloves
Ideal for use in foundries, glass industry, oven construction, etc.
Certifikace: EN 388 (2442), EN 407 (4443XX), Cat. III.