(±)-Naringenin

Supplier: Cayman Chemical

Synonyms: (±)-2,3-Dihydro-5,7-dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-2-(4-hydroxyphenyl)chroman-4-one, 4',5,7-Trihydroxyflavanone, Naringenin

14173-1 14173-5 14173-10
CAYM14173-1EA 50 EUR
CAYM14173-1 CAYM14173-5 CAYM14173-10
(±)-Naringenin
(±)-Naringenin
Formula: C₁₅H₁₂O₅
MW: 272.25 g/mol
MDL Number: MFCD00006844
CAS Number: 480-41-1
EINECS: 207-550-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR