5,6-Dichloro-2-methylbenzimidazole
Supplier: Thermo Scientific
TL00812.1
TL00812.10
MAYBTL00812.1EA
0
EUR
MAYBTL00812.1
MAYBTL00812.10
5,6-Dichloro-2-methylbenzimidazole
5,6-Dichloro-2-methylbenzimidazole
Formula:
C₈H₆Cl₂N₂ MW: 201.0500 g/mol |
MDL Number:
MFCD00276973 CAS Number: 6478-79-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...