Bis(cyclopentadienyl)hafnium dichloride, Sigma-Aldrich®
Supplier: Merck
Synonyms: Hafnocene dichloride
M-Clarity™ quality level = MQ200
119334-10G 119334-1G
SIAL119334-10GEA 356 EUR
SIAL119334-10G SIAL119334-1G
Bis(cyclopentadienyl)hafnium dichloride, Sigma-Aldrich®
Bis(cyclopentadienyl)hafnium dichloride
Formula: C₁₀H₁₀Cl₂Hf
MW: 379.58 g/mol
MDL Number: MFCD00001438
CAS Number: 12116-66-4
EINECS: 235-177-5