N,N-Dimethyl-2,3-dichloroaniline

Supplier: Thermo Scientific

SEW05704.1 SEW05704.10 SEW05704.50
MAYBSEW05704.1EA 0 EUR
MAYBSEW05704.1 MAYBSEW05704.10 MAYBSEW05704.50
N,N-Dimethyl-2,3-dichloroaniline
N,N-Dimethyl-2,3-dichloroaniline
Formula: C₈H₉Cl₂N
CAS Number: 58566-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR