1-Methylimidazole-2-carbaldehyde, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1-Methyl-1H-imidazole-2-carbaldehyde, 1-Methylimidazole-2-carboxaldehyde
475122-5G 475122-1G
SIAL475122-5GEA 160 EUR
SIAL475122-5G SIAL475122-1G
1-Methylimidazole-2-carbaldehyde, Sigma-Aldrich®
1-Methylimidazole-2-carbaldehyde
Formula: C₅H₆N₂O
MW: 110.12 g/mol
MDL Number: MFCD01321308
CAS Number: 13750-81-7