Picolinic acid N-oxide, Sigma-Aldrich®

Supplier: Merck

P43009-25G P43009-5G
SIALP43009-25GEA 117 EUR
SIALP43009-25G SIALP43009-5G
Picolinic acid N-oxide, Sigma-Aldrich®
Picolinic acid N-oxide
Formula: C₆H₅NO₃
MW: 139.1088 g/mol
MDL Number: MFCD00006198
CAS Number: 824-40-8
EINECS: 212-528-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR