1-Methyl-4-(tributylstannyl)imidazole, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
719501-500MG
719501-1G
719501-5G
SIAL719501-500MGEA
175
EUR
SIAL719501-500MG
SIAL719501-1G
SIAL719501-5G
1-Methyl-4-(tributylstannyl)imidazole, Sigma-Aldrich®
1-Methyl-4-(tributylstannyl)imidazole
Formula:
C₁₆H₃₂N₂Sn |
MDL Number:
MFCD13193061 CAS Number: 446285-73-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...