Ethyl-3-(2-furfurylthio)propionate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Ethyl 3-(furfurylthio)propionate

M-Clarity™ quality level = MQ300

W367400-100G-K W367400-1KG-K W367400-25G-K
SIALW367400-100G-KEA 180 EUR
SIALW367400-100G-K SIALW367400-1KG-K SIALW367400-25G-K
Ethyl-3-(2-furfurylthio)propionate, Sigma-Aldrich®
Ethyl 3-(2-furfurylthio)propionate
Formula: C₁₀H₁₄O₃S
MW: 214.29 g/mol
MDL Number: MFCD00051819
CAS Number: 94278-27-0
EINECS: 304-716-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR