1-(2-Morpholinoethyl)-1H-pyrazole-4-boronic acid pinacol ester, Sigma-Aldrich®
Supplier: Merck
Synonyms:
4-(2-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)ethyl)morpholine
M-Clarity™ quality level = MQ100
706213-1G
706213-250MG
SIAL706213-1GEA
477
EUR
SIAL706213-1G
SIAL706213-250MG
1-(2-Morpholinoethyl)-1H-pyrazole-4-boronic acid pinacol ester, Sigma-Aldrich®
1-(2-Morpholinoethyl)-1H-pyrazole-4-boronic acid pinacol ester
Formula:
C₁₅H₂₆BN₃O₃ MW: 307.1962 g/mol |
MDL Number:
MFCD08062342 CAS Number: 864754-18-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...