(S)-(+)-3-Fluoropyrrolidine hydrochloride ≥97%
Supplier: Thermo Scientific
Warning
Synonyms: (S)-3-Fluoropyrrolidine hydrochloride
H27536.03 H27536.MD
H27536.03EA 196 EUR
H27536.03 H27536.MD
(S)-(+)-3-Fluoropyrrolidine hydrochloride ≥97%
(S)-(+)-3-Fluoropyrrolidine hydrochloride
Purity: 97% CAS No.: 136725-53-6 Molecular Formula: C4H8FN.HCl Formula Weight: 125.57 Melting Point: 183-186° MDL No.: MFCD04038718
Formula: C₄H₉ClFN
MW: 125.57 g/mol
Melting Pt: 183…186 °C
Storage Temperature: Ambient
MDL Number: MFCD04038718
CAS Number: 136725-53-6