4-(Piperidin-1-ylsulphonyl)aniline

Supplier: Thermo Scientific

TL00817DA TL00817EA
MAYBTL00817DAEA 0 EUR
MAYBTL00817DA MAYBTL00817EA
4-(Piperidin-1-ylsulphonyl)aniline
4-(Piperidin-1-ylsulphonyl)aniline
Formula: C₁₁H₁₆N₂O₂S
MW: 240.320 g/mol
MDL Number: MFCD00276969
CAS Number: 6336-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR