1-(3-Chlorophenyl)piperazine hydrochloride
Supplier: LGC Standards PROMOCHEM
PROONMIAD907EA
265
EUR
PROONMIAD907
1-(3-Chlorophenyl)piperazine hydrochloride
1-(3-Chlorophenyl)piperazine hydrochloride
Formula:
C₁₀H₁₃ClN₂·xHCl MW: 196,68 g/mol (anhy) |
MDL Number:
MFCD00039032 CAS Number: 65369-76-8 EINECS: 265-718-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...