Ethyl-4-chloro-2-(methylthio)pyrimidine-5-carboxylate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Ethyl 4-chloro-2-methylthio-5-pyrimidinecarboxylate, Ethyl 4-chloro-2-(methylsulphanyl)pyrimidine-5-carboxylate, Ethyl 4-chloro-2-(methylthio)pyrimidine-5-carboxylate
M-Clarity™ quality level = MQ100
145963-100G 145963-25G 145963-5G
SIAL145963-100GEA 293 EUR
SIAL145963-100G SIAL145963-25G SIAL145963-5G
Ethyl-4-chloro-2-(methylthio)pyrimidine-5-carboxylate, Sigma-Aldrich®
Ethyl 4-chloro-2-(methylthio)pyrimidine-5-carboxylate
Formula: C₈H₉ClN₂O₂S
MW: 232.69 g/mol
MDL Number: MFCD00006085
CAS Number: 5909-24-0
EINECS: 227-619-0