4-Chloro-2-(methylthio)pyrimidine, Sigma-Aldrich®

Supplier: Merck

Synonyms: 4-Chloro-2-methylthiopyrimidine

M-Clarity™ quality level = MQ200

145289-10G 145289-50G
SIAL145289-10GEA 39.1 EUR
SIAL145289-10G SIAL145289-50G
4-Chloro-2-(methylthio)pyrimidine, Sigma-Aldrich®
4-Chloro-2-(methylthio)pyrimidine
Formula: C₅H₅ClN₂S
MW: 160.63 g/mol
MDL Number: MFCD00006083
CAS Number: 49844-90-8
EINECS: 256-500-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR