Bathophenanthrolinedisulphonic acid disodium salt hydrate, Sigma-Aldrich®
Supplier: Merck
Synonyms: 4,7-Diphenyl-1,10-phenanthrolinedisulphonic acid disodium salt hydrate
M-Clarity™ quality level = MQ100
146617-1G 146617-5G
SIAL146617-1GEA 225 EUR
SIAL146617-1G SIAL146617-5G
Bathophenanthrolinedisulphonic acid disodium salt hydrate, Sigma-Aldrich®
Bathophenanthrolinedisulphonic acid disodium salt hydrate
Formula: C₂₄H₁₄N₂Na₂O₆S₂·H₂O
MW: 536.48 g/mol (anhy)
MDL Number: MFCD00149310
CAS Number: 52746-49-3
EINECS: 258-152-0