1-Ethoxy-1-(tributylstannyl)ethene 95%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Tributyl(1-ethoxyvinyl)tin, Tri-n-butyl(1-ethoxyvinyl)tin, 1-Ethoxyvinyltri-n-butyltin
H31562.03 H31562.06 H31562.14
H31562.03EA 59.4 EUR
H31562.03 H31562.06 H31562.14
1-Ethoxy-1-(tributylstannyl)ethene 95%
1-Ethoxy-1-(tributylstannyl)ethene
CAS No.: 97674-02-7 Molecular Formula: C16H34OSn Formula Weight: 361.14 Flash Point: 110°(230°F) Refractive Index: 1.4760 Boiling Point: 85-86°/0.07mm UN No.: 2788 MDL No.: MFCD00010240
Formula: C₁₆H₃₄OSn
Boiling Pt: 85…86 °C (0.07 mmHg)
Density: 1.067
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00010240
CAS Number: 97674-02-7
UN: 2788
ADR: 6.1,III