cis-1-Ethoxy-2-(tributylstannyl)ethene ≥94%
Supplier: Thermo Fisher Scientific
Danger
H51258.03 H51258.06
H51258.03EA 99.1 EUR
H51258.03 H51258.06
cis-1-Ethoxy-2-(tributylstannyl)ethene ≥94%
cis-1-Ethoxy-2-(tributylstannyl)ethene
Purity: 94% CAS No.: 64724-29-4 Molecular Formula: C16H34OSn Formula Weight: 361.15 Melting Point: 84-87° UN No.: 2788 MDL No.: MFCD07787391
Formula: C₁₆H₃₄OSn
MW: 361.15 g/mol
Melting Pt: 84…87 °C
Density: 1.082
Storage Temperature: Ambient
MDL Number: MFCD07787391
CAS Number: 64724-29-4
ADR: 6.1 III