cis-1-Ethoxy-2-(tributylstannyl)ethene, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
707937-1G
707937-5G
SIAL707937-1GEA
102
EUR
SIAL707937-1G
SIAL707937-5G
cis-1-Ethoxy-2-(tributylstannyl)ethene, Sigma-Aldrich®
cis-1-Ethoxy-2-(tributylstannyl)ethene
Formula:
C₁₆H₃₄OSn MW: 361.15 g/mol |
MDL Number:
MFCD07787391 CAS Number: 64724-29-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...