N-Boc-L-alaninol, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

SIAL469513-5GEA 49.6 EUR
SIAL469513-5G
N-Boc-L-alaninol, Sigma-Aldrich®
N-Boc-L-alaninol
Formula: C₈H₁₇NO₃
MW: 175,2 g/mol
MDL Number: MFCD00043121
CAS Number: 79069-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR