N-Boc-L-tyrosine 99+%
Supplier: Thermo Scientific
Synonyms: BOC-L-Tyrosine, Boc-Tyr-OH
275700050 275700250 275701000
ACRO275700050EA 41.2 EUR
ACRO275700050 ACRO275700250 ACRO275701000
N-Boc-L-tyrosine 99+%
N-Boc-L-tyrosine
Formula: C₁₄H₁₉NO₅
MW: 281.3 g/mol
Melting Pt: 135…140 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00037179
CAS Number: 3978-80-1
EINECS: 223-613-7
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤1.0 % (K.F.)
HPLC ≥99.0 %
Specific optical rotation +36.5° to +39.5° (24°C, 589 nm) (c=1, dioxane)