2-Fluoro-α-methyl-4-biphenylacetic acid
Supplier: Cayman Chemical
Synonyms: Ansaid, 2-(2-Fluoro-[1,1'-biphenyl]-4-yl)propanoic acid, Flurbiprofen, Froben, Antadys
70250-1 70250-5 70250-10 70250-25
CAYM70250-1EA 43 EUR
CAYM70250-1 CAYM70250-5 CAYM70250-10 CAYM70250-25
2-Fluoro-α-methyl-4-biphenylacetic acid
2-Fluoro-α-methyl-4-biphenylacetic acid
Formula: C₁₅H₁₃FO₂
MW: 244.265 g/mol
MDL Number: MFCD00079303
CAS Number: 5104-49-4
EINECS: 225-827-6