4-Phenyl-3-butyn-2-one, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
161268-1G
161268-5G
SIAL161268-1GEA
100
EUR
SIAL161268-1G
SIAL161268-5G
4-Phenyl-3-butyn-2-one, Sigma-Aldrich®
4-Phenyl-3-butyn-2-one
Formula:
C₁₀H₈O MW: 144.17 g/mol |
MDL Number:
MFCD00008776 CAS Number: 1817-57-8 EINECS: 217-327-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...